Assay Details
| Gene Symbol: LOC4324988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760975.2 | XP_015616461.1 | 4 - 5 | 993 | 124 | |
| GenBank mRNA | AK072213.1 | - | 4 - 5 | 604 | 124 | |
| AK120875.1 | - | 4 - 5 | 1001 | 124 | ||
More Information
Set Membership: |
|

LOC4324988
Genomic Map