Assay Details
Gene Symbol: LOC4324328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769999.2 | XP_015625485.1 | 3 - 4 | 232 | 118 | |
GenBank mRNA | AK103859.1 | - | 3 - 4 | 211 | 118 | |
BT129192.1 | - | 3 - 4 | 217 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
