Assay Details
| Gene Symbol: LOC4327752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760584.2 | XP_015616070.1 | 3 - 4 | 497 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4327752| Gene Symbol: LOC4327752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760584.2 | XP_015616070.1 | 3 - 4 | 497 | 86 | |
Set Membership: |
|