Assay Details
| Gene Symbol: LOC4326959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795517.2 | XP_015651003.1 | 2 - 3 | 1024 | 90 | |
| GenBank mRNA | AK102594.1 | - | 3 - 4 | 939 | 90 | |
| HQ858878.1 | - | 2 - 3 | 890 | 90 | ||
More Information
Set Membership: |
|

LOC4326959
Genomic Map