Assay Details
Gene Symbol: LOC4326960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766701.2 | XP_015622187.1 | 15 - 16 | 1566 | 103 | |
GenBank mRNA | AK120714.1 | - | 15 - 16 | 1573 | 103 | |
BT129140.1 | - | 4 - 5 | 241 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
