Assay Details
Gene Symbol: LOC4326960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766701.2 | XP_015622187.1 | 1 - 2 | 487 | 84 | |
GenBank mRNA | AK120714.1 | - | 1 - 2 | 494 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
