Assay Details
Gene Symbol: LOC4326960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766701.2 | XP_015622187.1 | 8 - 9 | 1005 | 99 | |
GenBank mRNA | AK120714.1 | - | 8 - 9 | 1012 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
