Assay Details
| Gene Symbol: LOC4325249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767023.1 | XP_015622509.1 | 10 - 11 | 1248 | 126 | |
| XM_015767028.2 | XP_015622514.1 | 9 - 10 | 1142 | 126 | ||
More Information
Set Membership: |
|

LOC4325249
Genomic Map