Assay Details
| Gene Symbol: LOC4325249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767023.1 | XP_015622509.1 | 13 - 14 | 1537 | 67 | |
| XM_015767028.2 | XP_015622514.1 | 12 - 13 | 1431 | 67 | ||
| GenBank mRNA | AK105424.1 | - | 13 - 14 | 1544 | 67 | |
More Information
Set Membership: |
|

LOC4325249
Genomic Map