Assay Details
| Gene Symbol: LOC4325249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767023.1 | XP_015622509.1 | 3 - 4 | 546 | 74 | |
| XM_015767028.2 | XP_015622514.1 | 3 - 4 | 554 | 74 | ||
| GenBank mRNA | AK105424.1 | - | 3 - 4 | 552 | 74 | |
More Information
Set Membership: |
|

LOC4325249
Genomic Map