Assay Details
| Gene Symbol: LOC4324462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758919.2 | XP_015614405.1 | 1 - 2 | 539 | 150 | |
| GenBank mRNA | AK107433.1 | - | 1 - 2 | 548 | 150 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015758919.2 transcripts.
Set Membership: |
|

LOC4324462
Genomic Map