Assay Details
| Gene Symbol: LOC4326401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759488.2 | XP_015614974.1 | 7 - 8 | 770 | 93 | |
| GenBank mRNA | AK060011.1 | - | 7 - 8 | 753 | 93 | |
| AK068879.1 | - | 7 - 8 | 768 | 93 | ||
| AK099210.1 | - | 7 - 8 | 771 | 93 | ||
| AK104863.1 | - | 7 - 8 | 760 | 93 | ||
More Information
Set Membership: |
|

LOC4326401
Genomic Map