Assay Details
| Gene Symbol: LOC4326401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759488.2 | XP_015614974.1 | 9 - 10 | 1049 | 64 | |
| GenBank mRNA | AK060011.1 | - | 9 - 10 | 1032 | 64 | |
| AK068879.1 | - | 9 - 10 | 1047 | 64 | ||
| AK099210.1 | - | 9 - 10 | 1050 | 64 | ||
| AK104863.1 | - | 9 - 10 | 1039 | 64 | ||
More Information
Set Membership: |
|

LOC4326401
Genomic Map