Assay Details
Gene Symbol: LOC4327977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771951.2 | XP_015627437.1 | 9 - 10 | 1727 | 88 | |
XM_015771952.2 | XP_015627438.1 | 10 - 11 | 1697 | 88 | ||
GenBank mRNA | AK074023.1 | - | 10 - 11 | 1688 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
