Assay Details
| Gene Symbol: LOC4327977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771949.2 | XP_015627435.1 | 10 - 11 | 1775 | 101 | |
| XM_015771950.2 | XP_015627436.1 | 11 - 12 | 1745 | 101 | ||
| XM_015771951.2 | XP_015627437.1 | 10 - 11 | 1766 | 101 | ||
| XM_015771952.2 | XP_015627438.1 | 11 - 12 | 1736 | 101 | ||
| XM_015771953.2 | XP_015627439.1 | 9 - 10 | 1753 | 101 | ||
| GenBank mRNA | AK061115.1 | - | 5 - 6 | 737 | 101 | |
| AK074023.1 | - | 11 - 12 | 1727 | 101 | ||
More Information
Set Membership: |
|

LOC4327977
Genomic Map