Assay Details
Gene Symbol: LOC4327977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771949.2 | XP_015627435.1 | 11 - 12 | 1838 | 73 | |
XM_015771950.2 | XP_015627436.1 | 12 - 13 | 1808 | 73 | ||
XM_015771951.2 | XP_015627437.1 | 11 - 12 | 1829 | 73 | ||
XM_015771952.2 | XP_015627438.1 | 12 - 13 | 1799 | 73 | ||
XM_015771953.2 | XP_015627439.1 | 10 - 11 | 1816 | 73 | ||
GenBank mRNA | AK061115.1 | - | 6 - 7 | 800 | 73 | |
AK074023.1 | - | 12 - 13 | 1790 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
