Assay Details
Gene Symbol: LOC4327977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771949.2 | XP_015627435.1 | 1 - 2 | 445 | 67 | |
XM_015771950.2 | XP_015627436.1 | 2 - 3 | 415 | 67 | ||
XM_015771951.2 | XP_015627437.1 | 1 - 2 | 445 | 67 | ||
XM_015771952.2 | XP_015627438.1 | 2 - 3 | 415 | 67 | ||
XM_015771953.2 | XP_015627439.1 | 1 - 2 | 447 | 67 | ||
GenBank mRNA | AK061115.1 | - | 1 - 2 | 288 | 67 | |
AK074023.1 | - | 2 - 3 | 406 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
