Assay Details
| Gene Symbol: LOC4327977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771949.2 | XP_015627435.1 | 8 - 9 | 1589 | 75 | |
| XM_015771950.2 | XP_015627436.1 | 9 - 10 | 1559 | 75 | ||
| XM_015771951.2 | XP_015627437.1 | 8 - 9 | 1589 | 75 | ||
| XM_015771952.2 | XP_015627438.1 | 9 - 10 | 1559 | 75 | ||
| XM_015771953.2 | XP_015627439.1 | 7 - 8 | 1567 | 75 | ||
| GenBank mRNA | AK074023.1 | - | 9 - 10 | 1550 | 75 | |
More Information
Set Membership: |
|

LOC4327977
Genomic Map