Assay Details
| Gene Symbol: LOC4328120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769738.2 | XP_015625224.1 | 1 - 2 | 351 | 108 | |
| GenBank mRNA | AK070123.1 | - | 1 - 2 | 336 | 108 | |
| AK121751.1 | - | 1 - 2 | 357 | 108 | ||
More Information
Set Membership: |
|

LOC4328120
Genomic Map