Assay Details
| Gene Symbol: LOC4328227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771287.2 | XP_015626773.1 | 2 - 3 | 826 | 126 | |
| GenBank mRNA | AK100570.1 | - | 2 - 3 | 826 | 126 | |
More Information
Set Membership: |
|

LOC4328227
Genomic Map