Assay Details
| Gene Symbol: LOC4328265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771563.2 | XP_015627049.1 | 12 - 13 | 3309 | 115 | |
| XM_015771564.2 | XP_015627050.1 | 7 - 8 | 2544 | 115 | ||
More Information
Set Membership: |
|

LOC4328265
Genomic Map