Assay Details
| Gene Symbol: LOC4328265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771563.2 | XP_015627049.1 | 13 - 14 | 3492 | 95 | |
| XM_015771564.2 | XP_015627050.1 | 8 - 9 | 2727 | 95 | ||
More Information
Set Membership: |
|

LOC4328265
Genomic Map