Assay Details
| Gene Symbol: LOC4328294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770449.2 | XP_015625935.1 | 10 - 11 | 947 | 133 | |
| GenBank mRNA | AK059340.1 | - | 10 - 11 | 937 | 133 | |
| AK068065.1 | - | 6 - 7 | 465 | 133 | ||
| AK243199.1 | - | 10 - 11 | 935 | 133 | ||
| Gene Symbol: LOC4328289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770862.2 | XP_015626348.1 | 11 - 12 | 1008 | 133 | |
More Information
Set Membership: |
|

LOC4328294
Genomic Map