Assay Details
| Gene Symbol: LOC4328294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770449.2 | XP_015625935.1 | 11 - 12 | 1091 | 112 | |
| GenBank mRNA | AK059340.1 | - | 11 - 12 | 1081 | 112 | |
| AK068065.1 | - | 7 - 8 | 609 | 112 | ||
More Information
Set Membership: |
|

LOC4328294
Genomic Map