Assay Details
| Gene Symbol: LOC4328294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770449.2 | XP_015625935.1 | 3 - 4 | 472 | 119 | |
| GenBank mRNA | AK059340.1 | - | 3 - 4 | 462 | 119 | |
| AK243199.1 | - | 3 - 4 | 460 | 119 | ||
More Information
Set Membership: |
|

LOC4328294
Genomic Map