Assay Details
| Gene Symbol: LOC4328294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770449.2 | XP_015625935.1 | 9 - 10 | 869 | 117 | |
| GenBank mRNA | AK059340.1 | - | 9 - 10 | 859 | 117 | |
| AK068065.1 | - | 5 - 6 | 387 | 117 | ||
| AK243199.1 | - | 9 - 10 | 857 | 117 | ||
| Gene Symbol: LOC4328289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770862.2 | XP_015626348.1 | 10 - 11 | 930 | 117 | |
More Information
Set Membership: |
|

LOC4328294
Genomic Map