Assay Details
| Gene Symbol: LOC4341727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK288943.1 | - | 2413 | 65 | ||
| Gene Symbol: LOC4324727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK063441.1 | - | 79 | 65 | ||
| Gene Symbol: LOC4328390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769228.1 | XP_015624714.1 | 1 - 2 | 157 | 65 | |
| GenBank mRNA | AK061956.1 | - | 1 - 2 | 79 | 65 | |
| AK067024.1 | - | 1 - 2 | 82 | 65 | ||
| AK101547.1 | - | 1 - 2 | 82 | 65 | ||
| FJ940199.1 | - | 1 - 2 | 71 | 65 | ||
| GQ848039.1 | - | 1 - 2 | 51 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769228.1 transcripts.
Set Membership: |
|

LOC4341727
Genomic Map