Assay Details
| Gene Symbol: LOC4328498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768062.2 | XP_015623548.1 | 1 - 2 | 258 | 51 | |
| GenBank mRNA | AK058557.1 | - | 1 - 2 | 249 | 51 | |
More Information
Set Membership: |
|

LOC4328498
Genomic Map