Assay Details
| Gene Symbol: LOC4328498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768062.2 | XP_015623548.1 | 3 - 4 | 425 | 76 | |
| GenBank mRNA | AK058557.1 | - | 3 - 4 | 416 | 76 | |
More Information
Set Membership: |
|

LOC4328498
Genomic Map