Assay Details
| Gene Symbol: LOC4328498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768062.2 | XP_015623548.1 | 4 - 5 | 532 | 83 | |
| GenBank mRNA | AK058557.1 | - | 4 - 5 | 523 | 83 | |
More Information
Set Membership: |
|

LOC4328498
Genomic Map