Assay Details
| Gene Symbol: LOC4328593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769832.2 | XP_015625318.1 | 3 - 4 | 525 | 65 | |
| GenBank mRNA | AK060499.1 | - | 3 - 4 | 531 | 65 | |
More Information
Set Membership: |
|

LOC4328593
Genomic Map