Assay Details
| Gene Symbol: LOC4328593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769832.2 | XP_015625318.1 | 5 - 6 | 769 | 91 | |
| GenBank mRNA | AK060499.1 | - | 5 - 6 | 775 | 91 | |
More Information
Set Membership: |
|

LOC4328593
Genomic Map