Assay Details
| Gene Symbol: LOC4328601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768373.2 | XP_015623859.1 | 3 | 4404 | 91 | |
| GenBank mRNA | AK110011.1 | - | 1 | 1349 | 91 | |
More Information
Set Membership: |
|

LOC4328601
Genomic Map