Assay Details
| Gene Symbol: LOC4329033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772024.2 | XP_015627510.1 | 6 - 7 | 1291 | 90 | |
| XM_015772027.2 | XP_015627513.1 | 5 - 6 | 1001 | 90 | ||
| GenBank mRNA | AK064078.1 | - | 1 - 2 | 508 | 90 | |
More Information
Set Membership: |
|

LOC4329033
Genomic Map