Assay Details
| Gene Symbol: LOC4329226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767333.2 | XP_015622819.1 | 6 | 1306 | 144 | |
| XM_015767334.2 | XP_015622820.1 | 6 | 1321 | 144 | ||
| XM_015767335.2 | XP_015622821.1 | 6 | 1267 | 144 | ||
| XM_015767336.2 | XP_015622822.1 | 6 | 1367 | 144 | ||
| GenBank mRNA | AK065640.1 | - | 1 | 2044 | 144 | |
| AK288593.1 | - | 4290 | 144 | |||
More Information
Set Membership: |
|

LOC4329226
Genomic Map