Assay Details
| Gene Symbol: LOC4329538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769319.2 | XP_015624805.1 | 5 - 6 | 834 | 66 | |
| GenBank mRNA | AK069978.1 | - | 5 - 6 | 835 | 66 | |
More Information
Set Membership: |
|

LOC4329538
Genomic Map