Assay Details
| Gene Symbol: LOC4329597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771824.2 | XP_015627310.1 | 1 - 2 | 156 | 157 | |
| XM_015771825.2 | XP_015627311.1 | 1 - 2 | 156 | 157 | ||
| GenBank mRNA | AK066798.1 | - | 1 - 2 | 153 | 157 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771824.2, XM_015771825.2 transcripts.
Set Membership: |
|

LOC4329597
Genomic Map