Assay Details
| Gene Symbol: LOC4329597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771824.2 | XP_015627310.1 | 2 - 3 | 2365 | 116 | |
| XM_015771825.2 | XP_015627311.1 | 2 - 3 | 2365 | 116 | ||
| XM_015771826.2 | XP_015627312.1 | 3 - 4 | 2669 | 116 | ||
| XM_015771827.2 | XP_015627313.1 | 3 - 4 | 2731 | 116 | ||
| XM_015771828.2 | XP_015627314.1 | 3 - 4 | 2663 | 116 | ||
| XM_015771829.2 | XP_015627315.1 | 3 - 4 | 2725 | 116 | ||
| XM_015771831.2 | XP_015627317.1 | 3 - 4 | 2655 | 116 | ||
| XM_015771832.2 | XP_015627318.1 | 3 - 4 | 2717 | 116 | ||
| GenBank mRNA | AK066798.1 | - | 2 - 3 | 2362 | 116 | |
| AK121855.1 | - | 1 - 2 | 2745 | 116 | ||
More Information
Set Membership: |
|

LOC4329597
Genomic Map