Assay Details
| Gene Symbol: LOC4329817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771146.2 | XP_015626632.1 | 8 - 9 | 2265 | 60 | |
| GenBank mRNA | AK100170.1 | - | 8 - 9 | 2246 | 60 | |
| AK100542.1 | - | 8 - 9 | 2236 | 60 | ||
More Information
Set Membership: |
|

LOC4329817
Genomic Map