Assay Details
| Gene Symbol: LOC4329949 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772194.1 | XP_015627680.1 | 3 - 4 | 437 | 73 | |
| GenBank mRNA | AK102477.1 | - | 3 - 4 | 398 | 73 | |
More Information
Set Membership: |
|

LOC4329949
Genomic Map