Assay Details
| Gene Symbol: LOC4330525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768024.2 | XP_015623510.1 | 3 - 4 | 638 | 92 | |
| XM_015768025.1 | XP_015623511.1 | 4 - 5 | 608 | 92 | ||
| GenBank mRNA | AK287911.1 | - | 4 - 5 | 606 | 92 | |
More Information
Set Membership: |
|

LOC4330525
Genomic Map