Assay Details
| Gene Symbol: LOC4330582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769708.2 | XP_015625194.1 | 3 - 4 | 675 | 97 | |
| GenBank mRNA | AK058309.1 | - | 2 - 3 | 127 | 97 | |
| AK103602.1 | - | 3 - 4 | 681 | 97 | ||
More Information
Set Membership: |
|

LOC4330582
Genomic Map