Assay Details
| Gene Symbol: LOC4330649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770616.1 | XP_015626102.1 | 1 - 2 | 313 | 72 | |
| GenBank mRNA | AB037595.1 | - | 1 - 2 | 86 | 72 | |
| AK061157.1 | - | 1 - 2 | 168 | 72 | ||
| AK071969.1 | - | 1 - 2 | 319 | 72 | ||
| AK104987.2 | - | 1 - 2 | 134 | 72 | ||
| AK109397.2 | - | 1 - 2 | 134 | 72 | ||
| X14245.1 | - | 1 - 2 | 131 | 72 | ||
More Information
Set Membership: |
|

LOC4330649
Genomic Map