Assay Details
| Gene Symbol: LOC4330649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770616.1 | XP_015626102.1 | 6 - 7 | 695 | 64 | |
| GenBank mRNA | AB037595.1 | - | 6 - 7 | 468 | 64 | |
| AK061157.1 | - | 6 - 7 | 550 | 64 | ||
| AK071969.1 | - | 6 - 7 | 701 | 64 | ||
| AK104987.2 | - | 6 - 7 | 516 | 64 | ||
| AK109397.2 | - | 6 - 7 | 516 | 64 | ||
| X14245.1 | - | 6 - 7 | 513 | 64 | ||
More Information
Set Membership: |
|

LOC4330649
Genomic Map