Assay Details
| Gene Symbol: LOC4330649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770616.1 | XP_015626102.1 | 8 - 9 | 905 | 109 | |
| GenBank mRNA | AB037595.1 | - | 8 - 9 | 678 | 109 | |
| AK061157.1 | - | 8 - 9 | 760 | 109 | ||
| AK071969.1 | - | 8 - 9 | 911 | 109 | ||
| AK104987.2 | - | 8 - 9 | 726 | 109 | ||
| AK109397.2 | - | 8 - 9 | 726 | 109 | ||
| X14245.1 | - | 8 - 9 | 723 | 109 | ||
More Information
Set Membership: |
|

LOC4330649
Genomic Map