Assay Details
| Gene Symbol: LOC4330759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771878.1 | XP_015627364.1 | 4 - 5 | 825 | 129 | |
| GenBank mRNA | AK101787.1 | - | 4 - 5 | 766 | 129 | |
More Information
Set Membership: |
|

LOC4330759
Genomic Map