Assay Details
| Gene Symbol: LOC4330814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767556.2 | XP_015623042.1 | 2 | 757 | 67 | |
| XM_015767557.2 | XP_015623043.1 | 2 | 696 | 67 | ||
| XM_015767558.2 | XP_015623044.1 | 3 | 895 | 67 | ||
| GenBank mRNA | AK059923.1 | - | 1 | 1115 | 67 | |
| AK068833.1 | - | 1 | 1753 | 67 | ||
More Information
Set Membership: |
|

LOC4330814
Genomic Map