Assay Details
| Gene Symbol: LOC4330890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768767.2 | XP_015624253.1 | 2 - 3 | 482 | 113 | |
| GenBank mRNA | AK110707.1 | - | 2 - 3 | 482 | 113 | |
More Information
Set Membership: |
|

LOC4330890
Genomic Map