Assay Details
| Gene Symbol: LOC4330923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767464.2 | XP_015622950.1 | 19 - 20 | 1897 | 65 | |
| XM_015767465.2 | XP_015622951.1 | 19 - 20 | 1893 | 65 | ||
| GenBank mRNA | AK242973.1 | - | 19 - 20 | 1868 | 65 | |
More Information
Set Membership: |
|

LOC4330923
Genomic Map