Assay Details
| Gene Symbol: LOC4330923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767464.2 | XP_015622950.1 | 16 - 17 | 1569 | 102 | |
| XM_015767465.2 | XP_015622951.1 | 16 - 17 | 1565 | 102 | ||
| GenBank mRNA | AK242973.1 | - | 16 - 17 | 1540 | 102 | |
More Information
Set Membership: |
|

LOC4330923
Genomic Map